Assay Details
| Gene Symbol: Gprc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271106.1 | NP_001258035.1 | 4 - 5 | 1571 | 83 | |
| GenBank mRNA | AJ535460.1 | - | 4 - 5 | 1547 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gprc6a
Genomic Map