Assay Details
Gene Symbol: Sept10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014033.1 | NP_001014055.1 | 7 - 8 | 1068 | 94 | |
XM_006256415.2 | XP_006256477.1 | 8 - 9 | 1293 | 94 | ||
XM_006256417.2 | XP_006256479.1 | 7 - 8 | 1286 | 94 | ||
XM_006256418.2 | XP_006256480.1 | 8 - 9 | 1292 | 94 | ||
XM_008772886.1 | XP_008771108.1 | 7 - 8 | 1571 | 94 | ||
GenBank mRNA | BC087157.1 | - | 7 - 8 | 1068 | 7133379 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
