Assay Details
| Gene Symbol: Ddx50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013198.2 | NP_001013216.2 | 11 - 12 | 1689 | 151 | |
| GenBank mRNA | BC091427.1 | - | 5 - 6 | 575 | 7366491 | 151 |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx50
Genomic Map