Assay Details
| Gene Symbol: Tmem17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010961.1 | NP_001010961.1 | 2 - 3 | 547 | 110 | |
| GenBank mRNA | BC089059.1 | - | 2 - 3 | 547 | 7320241 | 110 |
| FQ228748.1 | - | 2 - 3 | 205 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem17
Genomic Map