Assay Details
| Gene Symbol: Acyp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169145.1 | NP_001162616.1 | 2 - 3 | 288 | 71 | |
| XM_008770465.1 | XP_008768687.1 | 2 - 3 | 386 | 71 | ||
| XM_008770466.1 | XP_008768688.1 | 2 - 3 | 369 | 71 | ||
| GenBank mRNA | CB613181.1 | - | 2 - 3 | 298 | 71 | |
| CO404339.1 | - | 2 - 3 | 119 | 71 | ||
| FQ215015.1 | - | 2 - 3 | 120 | 71 | ||
| FQ224926.1 | - | 2 - 3 | 156 | 71 | ||
More Information
Set Membership: |
|

Acyp2
Genomic Map