Assay Details
| Gene Symbol: Peli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100565.1 | NP_001094035.1 | 5 - 6 | 779 | 79 | |
| GenBank mRNA | BC166745.1 | - | 5 - 6 | 612 | 7939059 | 79 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Peli1
Genomic Map