Assay Details
Gene Symbol: Prl8a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134385.2 | NP_599212.1 | 2 - 3 | 247 | 130 | |
XM_006253904.1 | XP_006253966.1 | 2 - 3 | 238 | 130 | ||
GenBank mRNA | AF239745.1 | - | 2 - 3 | 247 | 130 | |
AF239746.1 | - | 2 - 3 | 244 | 130 | ||
AF239747.1 | - | 2 - 3 | 247 | 130 | ||
BC100138.1 | - | 2 - 3 | 240 | 7442617 | 130 | |
BC127474.1 | - | 2 - 3 | 238 | 7443385 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
