Assay Details
| Gene Symbol: Dusp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108412.1 | NP_001101882.1 | 5 - 6 | 387 | 69 | |
| XM_006253888.2 | XP_006253950.1 | 5 - 6 | 335 | 69 | ||
| XM_006253889.2 | XP_006253951.1 | 5 - 6 | 608 | 69 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dusp22
Genomic Map