Assay Details
| Gene Symbol: Dtnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037664.1 | NP_001032753.1 | 1 - 2 | 167 | 60 | |
| GenBank mRNA | BC088267.1 | - | 1 - 2 | 167 | 7379090 | 60 |
More Information
Updated Mapping Information
Set Membership: |
|

Dtnbp1
Genomic Map