Assay Details
Gene Symbol: Prl3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153736.1 | NP_714958.1 | 3 - 4 | 533 | 106 | |
GenBank mRNA | AB019791.1 | - | 3 - 4 | 535 | 106 | |
AF526270.1 | - | 3 - 4 | 533 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
