Assay Details
Gene Symbol: Ippk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008556.1 | NP_001008556.1 | 12 - 13 | 86 | ||
XM_008771534.1 | XP_008769756.1 | 11 - 12 | 1959 | 86 | ||
XM_008771535.1 | XP_008769757.1 | 12 - 13 | 1343 | 86 | ||
GenBank mRNA | AY823319.1 | - | 12 - 13 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
