Assay Details
| Gene Symbol: Slc22a23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022624.1 | NP_072146.1 | 4 - 5 | 1823 | 79 | |
| XM_006253845.2 | XP_006253907.1 | 4 - 5 | 1103 | 79 | ||
| GenBank mRNA | AF184921.1 | - | 1 - 2 | 91 | 79 | |
| BC158843.1 | - | 4 - 5 | 722 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a23
Genomic Map