Assay Details
| Gene Symbol: Prl8a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253907.1 | XP_006253969.1 | 2 - 3 | 262 | 103 | |
| XM_006253908.1 | XP_006253970.1 | 2 - 3 | 212 | 103 | ||
| GenBank mRNA | BC097274.1 | - | 1 - 2 | 199 | 7441946 | 103 |
| BC158577.1 | - | 2 - 3 | 246 | 7461869 | 103 | |
| M76537.1 | - | 2 - 3 | 229 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prl8a5
Genomic Map