Assay Details
| Gene Symbol: Serpinb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199085.2 | NP_954516.1 | 4 - 5 | 538 | 89 | |
| XM_006253869.1 | XP_006253931.1 | 4 - 5 | 536 | 89 | ||
| XM_006253870.2 | XP_006253932.1 | 4 - 5 | 831 | 89 | ||
| XM_006253871.2 | XP_006253933.1 | 4 - 5 | 651 | 89 | ||
| XM_008771596.1 | XP_008769818.1 | 4 - 5 | 749 | 89 | ||
| GenBank mRNA | BC060594.1 | - | 4 - 5 | 542 | 6887824 | 89 |
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb6
Genomic Map