Assay Details
| Gene Symbol: Rreb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107348.1 | NP_001100818.1 | 9 - 10 | 1235 | 69 | |
| XM_006253812.2 | XP_006253874.1 | 8 - 9 | 1213 | 69 | ||
| XM_006253813.2 | XP_006253875.1 | 7 - 8 | 967 | 69 | ||
| XM_006253814.2 | XP_006253876.1 | 8 - 9 | 1582 | 69 | ||
| XM_006253815.2 | XP_006253877.1 | 8 - 9 | 2179 | 69 | ||
| XM_006253816.2 | XP_006253878.1 | 8 - 9 | 1213 | 69 | ||
| XM_006253817.2 | XP_006253879.1 | 5 - 6 | 1712 | 69 | ||
| XM_008771571.1 | XP_008769793.1 | 7 - 8 | 967 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rreb1
Genomic Map