Assay Details
| Gene Symbol: Cks2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126083.1 | NP_001119555.1 | 1 - 2 | 165 | 89 | |
| GenBank mRNA | BC162005.1 | - | 1 - 2 | 165 | 9089459 | 89 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cks2
Genomic Map