Assay Details
| Gene Symbol: Tubb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013886.2 | NP_001013908.2 | 2 - 3 | 262 | 85 | |
| GenBank mRNA | BC105754.1 | - | 2 - 3 | 262 | 5599369 | 85 |
More Information
Updated Mapping Information
Set Membership: |
|

Tubb2b
Genomic Map