Assay Details
| Gene Symbol: LOC102554393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233879.2 | XP_006233941.2 | 27 - 28 | 3972 | 73 | |
| Gene Symbol: Cacfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106561.1 | NP_001100031.1 | 1 - 2 | 91 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106561.1 transcripts.
Set Membership: |
|

LOC102554393
Genomic Map