Assay Details
| Gene Symbol: Rsb66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181694.2 | NP_859045.2 | 2 - 3 | 226 | 104 | |
| XM_006233762.1 | XP_006233824.1 | 2 - 3 | 389 | 104 | ||
| XM_006233763.2 | XP_006233825.1 | 2 - 3 | 605 | 104 | ||
| XM_006233764.2 | XP_006233826.1 | 2 - 3 | 1060 | 104 | ||
| XM_006233766.1 | XP_006233828.1 | 2 - 3 | 389 | 104 | ||
| GenBank mRNA | BC097268.1 | - | 2 - 3 | 226 | 7457912 | 104 |
More Information
Updated Mapping Information
Set Membership: |
|

Rsb66
Genomic Map