Assay Details
| Gene Symbol: Ppp2r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108577.2 | NP_001102047.1 | 4 - 5 | 531 | 59 | |
| GenBank mRNA | BC166861.1 | - | 4 - 5 | 531 | 9101723 | 59 |
| FQ223957.1 | - | 5 - 6 | 686 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein peptidyl-prolyl isomerization
metabolic process
mitotic spindle organization in nucleus
negative regulation of phosphoprotein phosphatase activity
positive regulation of phosphoprotein phosphatase activity
positive regulation of protein dephosphorylation
negative regulation of protein dephosphorylation
positive regulation of apoptotic process
regulation of phosphoprotein phosphatase activity
peptidyl-prolyl cis-trans isomerase activity
receptor binding
ATP binding
protein tyrosine phosphatase activator activity
protein phosphatase type 2A regulator activity
ATPase activity
protein homodimerization activity
protein phosphatase 2A binding
protein peptidyl-prolyl isomerization
metabolic process
mitotic spindle organization in nucleus
negative regulation of phosphoprotein phosphatase activity
positive regulation of phosphoprotein phosphatase activity
positive regulation of protein dephosphorylation
negative regulation of protein dephosphorylation
positive regulation of apoptotic process
regulation of phosphoprotein phosphatase activity
peptidyl-prolyl cis-trans isomerase activity
receptor binding
ATP binding
protein tyrosine phosphatase activator activity
protein phosphatase type 2A regulator activity
ATPase activity
protein homodimerization activity
protein phosphatase 2A binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P047074 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P047068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166198 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047068 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047074 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047074 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047068 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P047068 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P047074 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P442161 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101489 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63326 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80977 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10835246 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10835248 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10835244 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10835243 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10835247 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10835251 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10835250 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10835249 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10835245 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10835242 | - | - |
| Rat Genome 230 2.0 Array | 1388338_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388338_at | - | - |
| Rat Genome 230 2.0 Array | 1395409_at | - | - |
| Rat Genome 230 2.0 Array | 1395409_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395409_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388338_at | - | - |
| Affymetrix HT Rat Focus Array | 1388338_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395409_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300002454.rn.1 | TC0300000212.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388338_PM_at | - | - |
| Rat Expression Set 230 | 1388338_at | - | - |
| Rat Expression Set 230 | 1395409_PM_at | - | - |
| Rat Expression Set 230 | 1395409_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10835242 | - | - |
| Rat Genome U34 Set | rc_AA849752_at | - | - |
| Rat Genome U34 Set | rc_AA945759_s_at | - | - |
| Rat Genome U34 Set | rc_AI012213_at | - | - |
| Rat Genome U34 Set | rc_AI231775_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22317187 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373881 | - | - |

Ppp2r4
Genomic Map