Assay Details
Gene Symbol: Uck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107831.3 | NP_001101301.2 | 1 - 2 | 195 | 78 | |
GenBank mRNA | BC168897.1 | - | 1 - 2 | 134 | 8362491 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
