Assay Details
Gene Symbol: Obp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128139.2 | NP_001121611.1 | 6 - 7 | 540 | 71 | |
GenBank mRNA | DQ537493.1 | - | 6 - 7 | 542 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
