Assay Details
| Gene Symbol: RGD1311084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100979.1 | NP_001094449.1 | 3 - 4 | 955 | 59 | |
| XM_006233913.2 | XP_006233975.1 | 4 - 5 | 929 | 59 | ||
| XM_006233914.2 | XP_006233976.1 | 4 - 5 | 929 | 59 | ||
| GenBank mRNA | BC127535.1 | - | 3 - 4 | 955 | 7133909 | 59 |
| FQ214464.1 | - | 3 - 4 | 700 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RGD1311084
Genomic Map