Assay Details
| Gene Symbol: RGD1311084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233913.2 | XP_006233975.1 | 1 - 2 | 632 | 100 | |
| XM_006233914.2 | XP_006233976.1 | 1 - 2 | 632 | 100 | ||
More Information
Set Membership: |
|

RGD1311084
Genomic Map