Assay Details
Gene Symbol: Triqk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171802.1 | NP_001165273.1 | 2 - 3 | 274 | 77 | |
GenBank mRNA | BC089993.1 | - | 2 - 3 | 244 | 7319652 | 77 |
CB805415.1 | - | 2 - 3 | 274 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC089993.1, NM_001171802.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
