Assay Details
| Gene Symbol: Pigo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238127.2 | XP_006238189.1 | 9 - 10 | 3181 | 72 | |
| XM_233141.8 | XP_233141.7 | 9 - 10 | 3724 | 72 | ||
More Information
Set Membership: |
|

Pigo
Genomic Map