Assay Details
Gene Symbol: Akirin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039914.2 | NP_001035003.1 | 2 - 3 | 576 | 101 | |
XM_008763614.1 | XP_008761836.1 | 2 - 3 | 823 | 101 | ||
GenBank mRNA | AB234867.1 | - | 2 - 3 | 596 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
