Assay Details
Gene Symbol: Slc26a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106638.1 | NP_001100108.1 | 17 - 18 | 1858 | 66 | |
XM_006237911.2 | XP_006237973.1 | 18 - 19 | 2270 | 66 | ||
XM_006237912.2 | XP_006237974.1 | 18 - 19 | 2178 | 66 | ||
XM_006237913.2 | XP_006237975.1 | 17 - 18 | 2081 | 66 | ||
XM_008763530.1 | XP_008761752.1 | 17 - 18 | 2193 | 66 | ||
XM_008763531.1 | XP_008761753.1 | 17 - 18 | 2105 | 66 | ||
XM_008763532.1 | XP_008761754.1 | 16 - 17 | 2097 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
gastric acid secretion
anion transport
chloride transport
sulfate transport
inorganic anion transport
bicarbonate transport
oxalate transport
ion transmembrane transport
sulfate transmembrane transport
chloride transmembrane transport
anion channel activity
chloride channel activity
secondary active sulfate transmembrane transporter activity
bicarbonate transmembrane transporter activity
sulfate transmembrane transporter activity
oxalate transmembrane transporter activity
gastric acid secretion
anion transport
chloride transport
sulfate transport
inorganic anion transport
bicarbonate transport
oxalate transport
ion transmembrane transport
sulfate transmembrane transport
chloride transmembrane transport
anion channel activity
chloride channel activity
secondary active sulfate transmembrane transporter activity
bicarbonate transmembrane transporter activity
sulfate transmembrane transporter activity
oxalate transmembrane transporter activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P398612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16600 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P398612 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P398612 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P398612 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P398612 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47277 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3491 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10875593 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875598 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875594 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875585 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875592 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875590 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875600 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875591 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875584 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875589 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875588 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875586 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875587 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875596 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875595 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875599 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875582 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10875581 | - | - |
Affymetrix Rat Clariom S Assay | PSR0500017678.rn.1 | TC0500002343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500017677.rn.1 | TC0500002343.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10875581 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21734449 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353252 | - | - |
