Assay Details
Gene Symbol: Ccnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100472.1 | NP_001093942.1 | 4 - 5 | 403 | 86 | |
XM_006237937.2 | XP_006237999.1 | 4 - 5 | 487 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cyclin-dependent protein serine/threonine kinase activity
transcription, DNA-templated
regulation of transcription, DNA-templated
protein ubiquitination
negative regulation of triglyceride metabolic process
protein kinase binding
ubiquitin protein ligase activity
regulation of cyclin-dependent protein serine/threonine kinase activity
transcription, DNA-templated
regulation of transcription, DNA-templated
protein ubiquitination
negative regulation of triglyceride metabolic process
protein kinase binding
ubiquitin protein ligase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P948572 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P373629 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P143840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114051 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P373629 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P948572 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P143840 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P948572 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P373629 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P143840 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P373629 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P143840 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P948572 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P475371 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P373629 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45972 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10867851 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867858 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867854 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867860 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867853 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867850 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867861 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867849 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867852 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867859 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867857 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867855 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867856 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10867848 | - | - |
Rat Genome 230 2.0 Array | 1390343_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389101_at | - | - |
Rat Genome 230 2.0 Array | 1390343_at | - | - |
Rat Genome 230 2.0 Array | 1389101_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390343_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389101_at | - | - |
Affymetrix HT Rat Focus Array | 1389101_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390343_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500001901.rn.1 | TC0500000309.rn.1 | RTA TC |
Rat Expression Set 230 | 1390343_PM_at | - | - |
Rat Expression Set 230 | 1390343_at | - | - |
Rat Expression Set 230 | 1389101_at | - | - |
Rat Expression Set 230 | 1389101_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867848 | - | - |
Rat Genome U34 Set | D14013_at | - | - |
Rat Genome U34 Set | rc_AA998893_at | - | - |
Rat Genome U34 Set | rc_AI232302_at | - | - |
Rat Genome U34 Set | D14013_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21350727 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371396 | - | - |
