Assay Details
Gene Symbol: Rad54b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191755.1 | NP_001178684.1 | 5 - 6 | 885 | 65 | |
XM_006237890.2 | XP_006237952.1 | 6 - 7 | 2690 | 65 | ||
XM_006237891.1 | XP_006237953.1 | 4 - 5 | 2738 | 65 | ||
XM_006237892.2 | XP_006237954.1 | 6 - 7 | 2690 | 65 | ||
XM_008763543.1 | XP_008761765.1 | 6 - 7 | 2690 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
