Assay Details
| Gene Symbol: Pdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237907.2 | XP_006237969.1 | 1 - 2 | 225 | 86 | |
| GenBank mRNA | BC129095.1 | - | 1 - 2 | 96 | 8369880 | 86 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC129095.1 transcripts.
Set Membership: |
|

Pdp1
Genomic Map