Assay Details
| Gene Symbol: Rragd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106641.2 | NP_001100111.1 | 5 - 6 | 994 | 73 | |
| GenBank mRNA | BC167014.1 | - | 5 - 6 | 994 | 9034554 | 73 |
More Information
Updated Mapping Information
Set Membership: |
|

Rragd
Genomic Map