Assay Details
| Gene Symbol: Icos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022610.1 | NP_072132.1 | 2 - 3 | 430 | 144 | |
| XM_006245037.2 | XP_006245099.1 | 4 - 5 | 682 | 144 | ||
| XM_006245038.2 | XP_006245100.1 | 4 - 5 | 679 | 144 | ||
| XM_008767136.1 | XP_008765358.1 | 4 - 5 | 697 | 144 | ||
| GenBank mRNA | AB023133.1 | - | 2 - 3 | 430 | 144 | |
| AB023134.1 | - | 2 - 3 | 430 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Icos
Genomic Map