Assay Details
Gene Symbol: Mitd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009714.1 | NP_001009714.1 | 624 | 96 | ||
XM_006244828.2 | XP_006244890.1 | 662 | 96 | |||
GenBank mRNA | BC088263.1 | - | 624 | 7376620 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
