Assay Details
| Gene Symbol: Ccdc115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108793.1 | NP_001102263.1 | 1 - 2 | 557 | 58 | |
| GenBank mRNA | BC166583.1 | - | 122 | 7442489 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc115
Genomic Map