Assay Details
Gene Symbol: Ccnyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767164.1 | XP_008765386.1 | 8 - 9 | 675 | 78 | |
XM_008767165.1 | XP_008765387.1 | 7 - 8 | 765 | 78 | ||
XM_008767166.1 | XP_008765388.1 | 8 - 9 | 674 | 78 | ||
XM_008767167.1 | XP_008765389.1 | 5 - 6 | 617 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
