Assay Details
| Gene Symbol: Satb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109306.1 | NP_001102776.1 | 3 - 4 | 627 | 67 | |
| XM_006244931.2 | XP_006244993.1 | 3 - 4 | 1299 | 67 | ||
| XM_008767128.1 | XP_008765350.1 | 2 - 3 | 1071 | 67 | ||
| XM_008767129.1 | XP_008765351.1 | 3 - 4 | 573 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Satb2
Genomic Map