Assay Details
Gene Symbol: Nif3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024763.1 | NP_001019934.1 | 1 - 2 | 539 | 60 | |
XM_006244958.2 | XP_006245020.1 | 3 - 4 | 680 | 60 | ||
XM_006244960.2 | XP_006245022.1 | 3 - 4 | 651 | 60 | ||
XM_006244961.2 | XP_006245023.1 | 3 - 4 | 676 | 60 | ||
XM_006244962.2 | XP_006245024.1 | 2 - 3 | 549 | 60 | ||
XM_006244963.2 | XP_006245025.1 | 3 - 4 | 683 | 60 | ||
XM_008767082.1 | XP_008765304.1 | 3 - 4 | 684 | 60 | ||
GenBank mRNA | BC097437.1 | - | 1 - 2 | 539 | 7455562 | 60 |
BC097992.1 | - | 3 - 4 | 612 | 7446502 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
