Assay Details
| Gene Symbol: Fcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031348.2 | NP_112638.2 | 3 - 4 | 355 | 88 | |
| XM_008761614.1 | XP_008759836.1 | 3 - 4 | 378 | 88 | ||
| GenBank mRNA | AB026057.2 | - | 3 - 4 | 266 | 88 | |
| BC088128.1 | - | 3 - 4 | 355 | 7376326 | 88 | |
| FQ227647.1 | - | 3 - 4 | 375 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fcna
Genomic Map