Assay Details
| Gene Symbol: Stkld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107822.1 | NP_001101292.1 | 3 - 4 | 195 | 63 | |
| XM_006233785.2 | XP_006233847.1 | 3 - 4 | 420 | 63 | ||
| XM_008761643.1 | XP_008759865.1 | 3 - 4 | 420 | 63 | ||
| XM_008761644.1 | XP_008759866.1 | 3 - 4 | 420 | 63 | ||
| XM_008761645.1 | XP_008759867.1 | 3 - 4 | 420 | 63 | ||
| GenBank mRNA | BC161886.1 | - | 3 - 4 | 192 | 7133966 | 63 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107822.1 transcripts.
Set Membership: |
|

Stkld1
Genomic Map