Assay Details
| Gene Symbol: Fcnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053634.1 | NP_446086.1 | 5 - 6 | 339 | 115 | |
| GenBank mRNA | AB036792.1 | - | 5 - 6 | 339 | 115 | |
| BC088123.1 | - | 5 - 6 | 331 | 7376834 | 115 | |
| FQ220557.1 | - | 5 - 6 | 342 | 115 | ||
| FQ233041.1 | - | 5 - 6 | 342 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fcnb
Genomic Map