Assay Details
| Gene Symbol: Ggta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145674.1 | NP_663707.1 | 5 - 6 | 528 | 128 | |
| XM_006234033.2 | XP_006234095.1 | 5 - 6 | 663 | 128 | ||
| XM_006234037.2 | XP_006234099.1 | 5 - 6 | 978 | 128 | ||
| XM_008761700.1 | XP_008759922.1 | 5 - 6 | 663 | 128 | ||
| XM_008761701.1 | XP_008759923.1 | 4 - 5 | 487 | 128 | ||
| XM_008761702.1 | XP_008759924.1 | 3 - 4 | 310 | 128 | ||
| GenBank mRNA | AF488784.1 | - | 4 - 5 | 285 | 128 | |
| AF520589.1 | - | 5 - 6 | 528 | 128 | ||
| BC166599.1 | - | 5 - 6 | 663 | 7932138 | 128 | |
| FQ222249.1 | - | 5 - 6 | 665 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ggta1
Genomic Map