Assay Details
| Gene Symbol: St6galnac6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015036.1 | NP_001015036.1 | 1 - 2 | 312 | 67 | |
| XM_006233982.2 | XP_006234044.1 | 2 - 3 | 755 | 67 | ||
| XM_006233984.2 | XP_006234046.1 | 3 - 4 | 922 | 67 | ||
| XM_006233985.2 | XP_006234047.1 | 2 - 3 | 753 | 67 | ||
| XM_006233987.2 | XP_006234049.1 | 3 - 4 | 387 | 67 | ||
| XM_006233988.2 | XP_006234050.1 | 3 - 4 | 387 | 67 | ||
| XM_008761770.1 | XP_008759992.1 | 4 - 5 | 599 | 67 | ||
| XM_008761771.1 | XP_008759993.1 | 4 - 5 | 598 | 67 | ||
| GenBank mRNA | AJ646881.1 | - | 2 - 3 | 196 | 67 | |
| BC091354.1 | - | 1 - 2 | 312 | 7317530 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC091354.1, NM_001015036.1 transcripts.
Set Membership: |
|

St6galnac6
Genomic Map