Assay Details
| Gene Symbol: St6galnac6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015036.1 | NP_001015036.1 | 2 - 3 | 721 | 79 | |
| XM_006233982.2 | XP_006234044.1 | 3 - 4 | 1164 | 79 | ||
| XM_006233984.2 | XP_006234046.1 | 4 - 5 | 1331 | 79 | ||
| XM_006233985.2 | XP_006234047.1 | 3 - 4 | 1162 | 79 | ||
| XM_006233987.2 | XP_006234049.1 | 4 - 5 | 796 | 79 | ||
| XM_006233988.2 | XP_006234050.1 | 4 - 5 | 796 | 79 | ||
| XM_008761770.1 | XP_008759992.1 | 5 - 6 | 1008 | 79 | ||
| XM_008761771.1 | XP_008759993.1 | 5 - 6 | 1007 | 79 | ||
| GenBank mRNA | AJ646881.1 | - | 3 - 4 | 605 | 79 | |
| BC091354.1 | - | 2 - 3 | 721 | 7317530 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

St6galnac6
Genomic Map