Assay Details
| Gene Symbol: Ccbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013164.3 | NP_001013182.3 | 11 - 12 | 1226 | 75 | |
| XM_006233810.2 | XP_006233872.1 | 11 - 12 | 1603 | 75 | ||
| XM_006233811.2 | XP_006233873.1 | 11 - 12 | 1696 | 75 | ||
| XM_006233812.2 | XP_006233874.1 | 11 - 12 | 1154 | 75 | ||
| XM_006233813.2 | XP_006233875.2 | 11 - 12 | 1166 | 75 | ||
| XM_008761658.1 | XP_008759880.1 | 12 - 13 | 1909 | 75 | ||
| GenBank mRNA | S61960.1 | - | 11 - 12 | 1367 | 75 | |
| Z49696.1 | - | 12 - 13 | 1375 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccbl1
Genomic Map