Assay Details
| Gene Symbol: Ccbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233811.2 | XP_006233873.1 | 1 - 2 | 567 | 91 | |
| GenBank mRNA | S61960.1 | - | 1 - 2 | 238 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the S61960.1, XM_006233811.2 transcripts.
Set Membership: |
|

Ccbl1
Genomic Map