Assay Details
Gene Symbol: Naa50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105881.1 | NP_001099351.1 | 561 | 118 | ||
XM_006248326.2 | XP_006248388.1 | 3 - 4 | 567 | 118 | ||
GenBank mRNA | BC169000.1 | - | 3 - 4 | 557 | 8362219 | 118 |
FQ216855.1 | - | 3 - 4 | 477 | 118 |
Gene Symbol: LOC102553099 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248358.2 | XP_006248420.1 | 3 - 4 | 320 | 118 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
