Assay Details
Gene Symbol: Stfa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105877.1 | NP_001099347.1 | 1 - 2 | 102 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
