Assay Details
| Gene Symbol: Fyttd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047899.2 | NP_001041364.1 | 4 - 5 | 467 | 150 | |
| XM_008768736.1 | XP_008766958.1 | 6 - 7 | 673 | 150 | ||
| XM_008768737.1 | XP_008766959.1 | 6 - 7 | 725 | 150 | ||
| GenBank mRNA | AY310146.1 | - | 4 - 5 | 507 | 150 | |
| AY724512.1 | - | 7 - 8 | 951 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P324180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78952 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P324180 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P324180 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P324180 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P553939 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P324180 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16871 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55587 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14965 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69224 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10754730 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754733 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754725 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754732 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754734 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754729 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754727 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754728 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754726 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754731 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10754722 | - | - |
| Rat Genome 230 2.0 Array | 1384945_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1384945_at | - | - |
| Rat Genome 230 2.0 Array | 1373836_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383098_at | - | - |
| Rat Genome 230 2.0 Array | 1383098_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373836_at | - | - |
| Affymetrix HT Rat Focus Array | 1383098_at | - | - |
| Affymetrix HT Rat Focus Array | 1373836_at | - | - |
| Affymetrix HT Rat Focus Array | 1384945_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373836_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1384945_at | - | - |
| Affymetrix HT Rat Focus Array | 1383098_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100010908.rn.1 | TC1100001744.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100010906.rn.1 | TC1100001744.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100010907.rn.1 | TC1100001744.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100010923.rn.1 | TC1100001744.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100010920.rn.1 | TC1100001744.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100010905.rn.1 | TC1100001744.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100010911.rn.1 | TC1100001744.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100010914.rn.1 | TC1100001744.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100010909.rn.1 | TC1100001744.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373836_PM_at | - | - |
| Rat Expression Set 230 | 1373836_at | - | - |
| Rat Expression Set 230 | 1383098_at | - | - |
| Rat Expression Set 230 | 1383098_PM_at | - | - |
| Rat Expression Set 230 | 1384945_PM_at | - | - |
| Rat Expression Set 230 | 1384945_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754722 | - | - |
| Rat Genome U34 Set | rc_AA801017_at | - | - |
| Rat Genome U34 Set | rc_AA925596_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21783336 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355027 | - | - |

Fyttd1
Genomic Map