Assay Details
| Gene Symbol: Igsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013120.1 | NP_001013138.1 | 4 - 5 | 766 | 104 | |
| GenBank mRNA | BC085929.1 | - | 4 - 5 | 766 | 7110445 | 104 |
More Information
Updated Mapping Information
Set Membership: |
|

Igsf11
Genomic Map